siRNA Details
Gene Symbol: Sec23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105732.2 | NP_001099202.1 | 17 | 2068 | ||
XM_006240121.2 | XP_006240183.1 | 17 | 2172 | |||
XM_008764661.1 | XP_008762883.1 | 17 | 2180 | |||
XM_008764662.1 | XP_008762884.1 | 17 | 2176 | |||
XM_008764663.1 | XP_008762885.1 | 16 | 2000 | |||
GenBank mRNA | BC169006.1 | - | 17 | 2068 | 7189145 |
