siRNA Details
Gene Symbol: Csnk1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031617.1 | NP_113805.1 | 4 | 432 | ||
XM_006242038.2 | XP_006242100.1 | 5 | 581 | |||
GenBank mRNA | AB042191.1 | - | 4 | 432 | ||
AB042192.1 | - | 4 | 432 | |||
AB056113.1 | - | 4 | 432 |
