siRNA Details
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031620.1 | NP_113808.1 | 2, 3 | 252 | ||
XM_006233010.1 | XP_006233072.1 | 1, 2 | 275 | |||
GenBank mRNA | BC086327.1 | - | 1, 2 | 280 | 7115765 | |
X97772.1 | - | 1, 2 | 211 |
