siRNA Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031628.1 | NP_113816.1 | 4, 5 | 1790 | ||
XM_008763702.1 | XP_008761924.1 | 5, 6 | 3584 | |||
XM_008763703.1 | XP_008761925.1 | 4, 5 | 3512 | |||
GenBank mRNA | D38530.1 | - | 4, 5 | 1790 | ||
DQ268830.1 | - | 4, 5 | 1790 | |||
X86003.1 | - | 2, 3 | 1577 |
