siRNA Details
Gene Symbol: Prl3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031316.1 | NP_112606.1 | 4 | 472 | ||
GenBank mRNA | AB019119.1 | - | 4 | 472 | ||
AB019945.1 | - | 4 | 463 | |||
AF150741.1 | - | 4 | 472 | |||
AF234638.1 | - | 4 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
