siRNA Details
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105733.1 | NP_001099203.1 | 4 | 645 | ||
XM_006234173.2 | XP_006234235.1 | 5 | 538 | |||
XM_008761773.1 | XP_008759995.1 | 4 | 4500 | |||
GenBank mRNA | L02315.1 | - | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
