siRNA Details
Gene Symbol: Nup210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053322.2 | NP_445774.1 | 9 | 1137 | ||
XM_006236921.2 | XP_006236983.1 | 7 | 1318 | |||
GenBank mRNA | Y00826.1 | - | 9 | 1138 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nup210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053322.2 | NP_445774.1 | 9 | 1137 | ||
XM_006236921.2 | XP_006236983.1 | 7 | 1318 | |||
GenBank mRNA | Y00826.1 | - | 9 | 1138 |