siRNA Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247542.2 | XP_006247604.1 | 11 | 2367 | ||
GenBank mRNA | EU012436.1 | - | 12 | 1093 | ||
EU012437.1 | - | 11 | 942 | |||
EU012438.1 | - | 12 | 1161 | |||
L24096.1 | - | 12 | 1174 | |||
L25438.1 | - | 12 | 1096 |
