siRNA Details
Gene Symbol: Degs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053323.2 | NP_445775.2 | 2 | 554 | ||
XM_006250403.2 | XP_006250465.1 | 2 | 849 | |||
XM_006250404.2 | XP_006250466.1 | 2 | 795 | |||
GenBank mRNA | AJ938081.1 | - | 2 | 507 | ||
AY036902.1 | - | 2 | 536 | |||
BC083727.1 | - | 2 | 554 | 7191263 | ||
FQ225269.1 | - | 2 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
