siRNA Details
Gene Symbol: Degs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY036902.1 | - | 2 | 229 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007853, AK149811, Y08460, AK002617, AK077257, and others.
