siRNA Details
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031648.2 | NP_113836.2 | 5 | 269 | ||
XM_006228838.2 | XP_006228900.1 | 6 | 489 | |||
XM_006228839.2 | XP_006228901.1 | 5 | 264 | |||
XM_008759184.1 | XP_008757406.1 | 5 | 278 | |||
GenBank mRNA | CB712570.1 | - | 5 | 269 | ||
FM109156.1 | - | 5 | 223 | |||
FQ211349.1 | - | 5 | 280 | |||
FQ224131.1 | - | 5 | 232 | |||
FQ224338.1 | - | 5 | 315 | |||
U72246.1 | - | 4 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
