siRNA Details
Gene Symbol: Slc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022853.2 | NP_074044.1 | 2 | 1431 | ||
GenBank mRNA | U17133.1 | - | 2 | 1432 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022853.2 | NP_074044.1 | 2 | 1431 | ||
GenBank mRNA | U17133.1 | - | 2 | 1432 |