siRNA Details
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031651.1 | NP_113839.1 | 13 | 1385 | ||
GenBank mRNA | L19102.1 | - | 13 | 1385 | ||
U08031.1 | - | 1369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
