siRNA Details
Gene Symbol: Grk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030829.1 | NP_110456.1 | 14 | 1673 | ||
XM_006231686.1 | XP_006231748.1 | 14 | 1726 | |||
GenBank mRNA | BC081792.1 | - | 14 | 1726 | 7123515 | |
U34841.1 | - | 14 | 1673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
