siRNA Details
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112712.1 | NP_001106183.1 | 6 | 507 | ||
NM_001112713.1 | NP_001106184.1 | 6 | 507 | |||
NM_031657.3 | NP_113845.2 | 6 | 507 | |||
XM_006253653.2 | XP_006253715.1 | 6 | 616 | |||
XM_008771504.1 | XP_008769726.1 | 5 | 774 | |||
GenBank mRNA | AF040750.1 | - | 6 | 507 | ||
EX488847.1 | - | 6 | 667 | |||
X97439.1 | - | 6 | 507 | |||
X97440.1 | - | 6 | 507 | |||
Y09365.1 | - | 6 | 535 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001038018, NM_001286064, NM_001286063, NM_001286066, NM_001112711, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
