siRNA Details
Gene Symbol: Grk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022928.1 | NP_075217.1 | 13 | 1532 | ||
XM_006251359.1 | XP_006251421.1 | 12 | 1275 | |||
XM_006251360.1 | XP_006251422.1 | 13 | 1368 | |||
XM_008770353.1 | XP_008768575.1 | 13 | 1368 | |||
XM_008770354.1 | XP_008768576.1 | 12 | 1275 | |||
XM_008770355.1 | XP_008768577.1 | 13 | 1368 | |||
XM_008770356.1 | XP_008768578.1 | 9 | 879 | |||
XM_008770357.1 | XP_008768579.1 | 8 | 923 | |||
XM_008770358.1 | XP_008768580.1 | 6 | 1477 | |||
GenBank mRNA | X97568.1 | - | 13 | 1532 |
