siRNA Details
Gene Symbol: Tgfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021578.2 | NP_067589.1 | 1 | 485 | ||
GenBank mRNA | AY550025.1 | - | 1 | 340 | ||
BC076380.1 | - | 1 | 485 | 7107411 | ||
X52498.1 | - | 1 | 752 |

More Information
Other Species Targeted

Gene Ontology Categories:
