siRNA Details
Gene Symbol: Nxf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021579.1 | NP_067590.1 | 8 | 802 | ||
GenBank mRNA | AF093139.1 | - | 8 | 854 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nxf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021579.1 | NP_067590.1 | 8 | 802 | ||
GenBank mRNA | AF093139.1 | - | 8 | 854 |