siRNA Details
Gene Symbol: Ninj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021595.2 | NP_067606.1 | 3 | 487 | ||
XM_008763255.1 | XP_008761477.1 | 3 | 997 | |||
XM_008763256.1 | XP_008761478.1 | 4 | 914 | |||
XM_008763257.1 | XP_008761479.1 | 4 | 673 | |||
XM_008763258.1 | XP_008761480.1 | 4 | 824 | |||
XM_008763259.1 | XP_008761481.1 | 3 | 633 | |||
GenBank mRNA | AB040815.1 | - | 3 | 488 | ||
AF250322.1 | - | 3 | 488 |
