siRNA Details
Gene Symbol: Ip6k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021660.2 | NP_067692.2 | 3 | 356 | ||
XM_008766564.1 | XP_008764786.1 | 2 | 322 | |||
XM_008766566.1 | XP_008764788.1 | 2 | 322 | |||
GenBank mRNA | BC083574.1 | - | 3 | 356 | 7112777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
