siRNA Details
Gene Symbol: Dnase2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021664.1 | NP_067696.1 | 6 | 1160 | ||
XM_006233464.2 | XP_006233526.1 | 7 | 1237 | |||
XM_006233465.2 | XP_006233527.1 | 7 | 1264 | |||
XM_006233466.2 | XP_006233528.1 | 7 | 1267 | |||
XM_006233468.2 | XP_006233530.1 | 7 | 1648 | |||
XM_008761521.1 | XP_008759743.1 | 7 | 1370 | |||
GenBank mRNA | AF178974.1 | - | 6 | 1160 |
