siRNA Details
Gene Symbol: Negr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021682.1 | NP_067714.1 | 4 | 631 | ||
GenBank mRNA | AB017139.1 | - | 4 | 631 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177274, NM_001039094, AK142298, AK035218, BC146007, and others.
Gene Ontology Categories:
