siRNA Details
Gene Symbol: Kcnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021688.3 | NP_067720.1 | 3 | 867 | ||
XM_006255820.2 | XP_006255882.1 | 3 | 1084 | |||
GenBank mRNA | AF022819.1 | - | 3 | 889 | ||
BC061807.1 | - | 3 | 867 | 5598415 |
