siRNA Details
Gene Symbol: Twist2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021691.2 | NP_067723.1 | 2 | 565 | ||
GenBank mRNA | BF550979.1 | - | 2 | 258 | ||
Y08139.1 | - | 2 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
