siRNA Details
Gene Symbol: Serpinb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021696.2 | NP_067728.1 | 5 | 676 | ||
XM_006249639.2 | XP_006249701.1 | 6 | 862 | |||
XM_008769495.1 | XP_008767717.1 | 6 | 759 | |||
GenBank mRNA | X64563.1 | - | 5 | 677 |
