siRNA Details
Gene Symbol: Tgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031659.1 | NP_113847.1 | 3 | 538 | ||
XM_006252014.2 | XP_006252076.1 | 3 | 2085 | |||
XM_008770691.1 | XP_008768913.1 | 3 | 2176 | |||
GenBank mRNA | BC097305.1 | - | 3 | 494 | 7462186 | |
FQ210559.1 | - | 3 | 543 | |||
M57263.1 | - | 3 | 538 |
