siRNA Details
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031662.1 | NP_113850.1 | 4, 5 | 567 | ||
XM_006246799.2 | XP_006246861.1 | 5, 6 | 767 | |||
XM_006246801.2 | XP_006246863.1 | 5, 6 | 714 | |||
XM_008767866.1 | XP_008766088.1 | 6, 7 | 856 | |||
GenBank mRNA | AB023658.1 | - | 4, 5 | 567 | ||
L42810.1 | - | 4, 5 | 567 | |||
S83194.1 | - | 4, 5 | 567 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_018883, BC017529, AF117384, AK144483.
Gene Ontology Categories:
