siRNA Details
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021741.2 | NP_068509.2 | 8, 9 | 1087 | ||
XM_006231136.2 | XP_006231198.1 | 8, 9 | 1202 | |||
GenBank mRNA | FQ222521.1 | - | 6, 7 | 952 | ||
X99330.1 | - | 8, 9 | 875 |
