siRNA Details
Gene Symbol: Cd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021744.1 | NP_068512.1 | 2 | 598 | ||
XM_006254603.2 | XP_006254665.1 | 2 | 930 | |||
GenBank mRNA | AF087943.1 | - | 2 | 598 | ||
BC061733.1 | - | 2 | 736 | 5598794 | ||
FQ219480.1 | - | 2 | 631 | |||
U51804.1 | - | 2 | 617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
