siRNA Details
Gene Symbol: Nr1h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021745.1 | NP_068513.1 | 9 | 1261 | ||
XM_008765260.1 | XP_008763482.1 | 8 | 1631 | |||
XM_008765261.1 | XP_008763483.1 | 8 | 1618 | |||
XM_008765262.1 | XP_008763484.1 | 9 | 1312 | |||
GenBank mRNA | BC083815.1 | - | 8 | 1139 | 7110420 | |
U18374.1 | - | 9 | 1261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
