siRNA Details
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021746.1 | NP_068514.1 | 11 | 1160 | ||
XM_008765781.1 | XP_008764003.1 | 11 | 889 | |||
GenBank mRNA | BC081950.1 | - | 10 | 1209 | ||
BC127549.1 | - | 3 | 387 | |||
X96488.1 | - | 11 | 1160 |
