siRNA Details
Gene Symbol: Csad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134454.1 | NP_001127926.1 | 2 | 250 | ||
NM_021750.2 | NP_068518.1 | 3 | 261 | |||
XM_006242438.2 | XP_006242500.1 | 3 | 670 | |||
XM_006242439.1 | XP_006242501.1 | 3 | 267 | |||
XM_006242440.2 | XP_006242502.1 | 4 | 746 | |||
XM_006242441.2 | XP_006242503.1 | 3 | 328 | |||
XM_006242442.2 | XP_006242504.1 | 4 | 361 | |||
XM_008765782.1 | XP_008764004.1 | 3 | 448 | |||
GenBank mRNA | AJ132661.1 | - | 4 | 366 | ||
BC081804.1 | - | 3 | 261 | 7106840 | ||
EU786150.1 | - | 2 | 233 | |||
EU906909.1 | - | 2 | 250 | |||
M64755.1 | - | 3 | 300 | |||
U74492.1 | - | 1 | 111 | |||
X94152.1 | - | 3 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
