siRNA Details
Gene Symbol: Prom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110137.1 | NP_001103607.1 | 12 | 1671 | ||
NM_021751.2 | NP_068519.2 | 13 | 1738 | |||
XM_008770211.1 | XP_008768433.1 | 14 | 1806 | |||
XM_008770212.1 | XP_008768434.1 | 14 | 2102 | |||
XM_008770213.1 | XP_008768435.1 | 14 | 1998 | |||
XM_008770214.1 | XP_008768436.1 | 14 | 1806 | |||
XM_008770215.1 | XP_008768437.1 | 13 | 2074 | |||
XM_008770216.1 | XP_008768438.1 | 14 | 1758 | |||
XM_008770217.1 | XP_008768439.1 | 14 | 1806 | |||
XM_008770218.1 | XP_008768440.1 | 13 | 1779 | |||
GenBank mRNA | AF263368.1 | - | 12 | 1705 | ||
AF386758.1 | - | 12 | 1617 | |||
AY262731.1 | - | 12 | 1617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
