siRNA Details
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021764.1 | NP_068532.2 | 11 | 1649 | ||
XM_006235252.2 | XP_006235314.1 | 12 | 1574 | |||
GenBank mRNA | AB011369.1 | - | 10 | 1488 | ||
BC061723.1 | - | 11 | 1583 | 5598480 | ||
U48248.1 | - | 11 | 1649 |

More Information
Other Species Targeted

Gene Ontology Categories:
