siRNA Details
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021772.2 | NP_068540.1 | 7 | 969 | ||
XM_006246323.2 | XP_006246385.1 | 7 | 1160 | |||
XM_006246325.2 | XP_006246387.1 | 7 | 1066 | |||
XM_006246326.2 | XP_006246388.1 | 7 | 1069 | |||
XM_006246327.2 | XP_006246389.1 | 7 | 1145 | |||
XM_006246329.2 | XP_006246391.1 | 7 | 1162 | |||
XM_006246332.2 | XP_006246394.1 | 7 | 1158 | |||
XM_006246333.2 | XP_006246395.1 | 7 | 1157 | |||
XM_008767705.1 | XP_008765927.1 | 7 | 1159 | |||
XM_008767706.1 | XP_008765928.1 | 7 | 1158 | |||
XM_008767707.1 | XP_008765929.1 | 7 | 1157 | |||
GenBank mRNA | AF112183.1 | - | 7 | 969 | ||
AF112184.1 | - | 7 | 905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
