siRNA Details
Gene Symbol: Fhit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021774.1 | NP_068542.1 | 5, 6 | 496 | ||
GenBank mRNA | AF170064.1 | - | 5, 6 | 496 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
