siRNA Details
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031664.1 | NP_113852.1 | 4 | 498 | ||
GenBank mRNA | AY029302.1 | - | 4 | 479 | ||
EU032627.2 | - | 4 | 566 | |||
U25055.1 | - | 4 | 498 | |||
U66723.1 | - | 4 | 469 |

More Information
Other Species Targeted

Gene Ontology Categories:
