siRNA Details
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021843.4 | NP_068615.1 | 3, 4 | 374 | ||
NM_021844.2 | NP_068616.1 | 3, 4 | 374 | |||
XM_008765339.1 | XP_008763561.1 | 3, 4 | 418 | |||
XM_008765340.1 | XP_008763562.1 | 3, 4 | 418 | |||
GenBank mRNA | AB105879.1 | - | 3, 4 | 374 | ||
AF071204.1 | - | 3, 4 | 209 | |||
AF071205.1 | - | 3, 4 | 210 | |||
CB751462.1 | - | 3, 4 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
