siRNA Details
Gene Symbol: Mcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021846.2 | NP_068618.2 | 736 | |||
GenBank mRNA | AF115380.1 | - | 668 | |||
BC078835.1 | - | 736 | 7121699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
