siRNA Details
Gene Symbol: Vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021847.3 | NP_068619.1 | 4 | 333 | ||
XM_006235706.2 | XP_006235768.2 | 4 | 507 | |||
GenBank mRNA | AF086631.1 | - | 4 | 333 | ||
BC065576.1 | - | 4 | 518 | 5598396 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019806, AF115504, AK081908, AK050632, BC057123, and others.
Gene Ontology Categories:
