siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021853.1 | NP_068625.1 | 30 | 3490 | ||
XM_006233697.2 | XP_006233759.1 | 30 | 3837 | |||
XM_006233698.2 | XP_006233760.1 | 30 | 3831 | |||
XM_006233699.2 | XP_006233761.1 | 30 | 3804 | |||
XM_006233700.2 | XP_006233762.1 | 29 | 3795 | |||
XM_006233702.2 | XP_006233764.1 | 29 | 3762 | |||
XM_006233704.2 | XP_006233766.1 | 29 | 3717 | |||
XM_006233705.2 | XP_006233767.1 | 29 | 3675 | |||
XM_006233706.2 | XP_006233768.1 | 30 | 3837 | |||
XM_008761624.1 | XP_008759846.1 | 30 | 3794 | |||
GenBank mRNA | AF089730.1 | - | 30 | 3490 | ||
AY884213.1 | - | 29 | 3388 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001145403, NM_175462, AK039085, AK048406, FJ628361, and others.
Gene Ontology Categories:
