siRNA Details
Gene Symbol: Tsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021854.1 | NP_068626.1 | 9 | 1107 | ||
XM_006233843.2 | XP_006233905.1 | 11 | 1561 | |||
XM_006233844.2 | XP_006233906.1 | 11 | 1532 | |||
XM_006233845.2 | XP_006233907.1 | 11 | 1383 | |||
XM_006233846.2 | XP_006233908.1 | 11 | 3648 | |||
XM_006233847.2 | XP_006233909.1 | 11 | 1869 | |||
XM_006233849.2 | XP_006233911.1 | 11 | 1561 | |||
GenBank mRNA | AB011821.1 | - | 9 | 1107 |
