siRNA Details
Gene Symbol: Gnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021858.3 | NP_068630.1 | 8, 9 | 802 | ||
GenBank mRNA | BC086422.1 | - | 8, 9 | 805 | 7193131 | |
DQ120487.1 | - | 6, 7 | 612 | |||
DQ120488.1 | - | 6, 7 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
