siRNA Details
Gene Symbol: Batf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021865.1 | NP_068637.1 | 3 | 310 | ||
XM_008769873.1 | XP_008768095.1 | 3 | 422 | |||
XM_008769874.1 | XP_008768096.1 | 3 | 450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
