siRNA Details
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021869.2 | NP_068641.2 | 4 | 292 | ||
XM_006227691.2 | XP_006227753.1 | 5 | 1614 | |||
XM_008758626.1 | XP_008756848.1 | 5 | 1608 | |||
GenBank mRNA | AF031430.1 | - | 3 | 196 | ||
BC085737.1 | - | 4 | 292 | 7193349 |
Gene Symbol: LOC100910446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758670.1 | XP_008756892.1 | 2 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
