siRNA Details
Gene Symbol: Il1rn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022194.2 | NP_071530.1 | 2 | 177 | ||
XM_006233636.2 | XP_006233698.1 | 3 | 2420 | |||
XM_006233637.2 | XP_006233699.1 | 3 | 207 | |||
XM_006233638.2 | XP_006233700.1 | 2 | 107 | |||
GenBank mRNA | BC070930.1 | - | 2 | 171 | ||
CK364274.1 | - | 2 | 201 |

More Information
Other Species Targeted

Gene Ontology Categories:
