siRNA Details
Gene Symbol: Clcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022198.2 | NP_071534.2 | 4 | 483 | ||
XM_006256828.2 | XP_006256890.1 | 4 | 786 | |||
GenBank mRNA | BC092209.1 | - | 4 | 483 | 7315063 | |
FQ210965.1 | - | 4 | 522 | |||
Z36944.1 | - | 2 | 191 |
