siRNA Details
Gene Symbol: Grm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022202.1 | NP_071538.1 | 8 | 1575 | ||
GenBank mRNA | U63288.1 | - | 8 | 1575 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022202.1 | NP_071538.1 | 8 | 1575 | ||
GenBank mRNA | U63288.1 | - | 8 | 1575 |
Set Membership: |
![]() |