siRNA Details
Gene Symbol: LOC60591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022203.1 | NP_071539.1 | 3 | 412 | ||
GenBank mRNA | U79661.1 | - | 3 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
