siRNA Details
Gene Symbol: Unc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022207.1 | NP_071543.1 | 14 | 2232 | ||
GenBank mRNA | U87306.1 | - | 14 | 2232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
